

5-Chloropyridine-2-sulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:885277-08-7
:
Formula:C₅H₃Cl₂NO₂S
MDL Number:MFCD06739113
:212.054 g/mol
UN:3261
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together