Order Entry
Netherlands
ContactUsLinkComponent
1-(2-Bromoethyl)-1H-pyrazole 95%
1-(2-Bromoethyl)-1H-pyrazole 95%
Catalog # APOSOR924221-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Bromoethyl)-1H-pyrazole 95%
Catalog # APOSOR924221-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.026 g/mol
Storage Temperature:Ambient
CAS Number:119291-22-4
Formula:C₅H₇BrN₂
MDL Number:MFCD03727235