2-Cyclobutylethan-1-amine
Catalog # APOSOR922462-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:99.174 g/mol
Storage Temperature:Ambient
CAS Number:60637-97-0
Formula:C₆H₁₃N
MDL Number:MFCD09924416