N,1-Dimethyl-1H-pyrazol-4-amine dihydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:948572-94-9
:
Formula:C₅H₉N₃·2ClH
MDL Number:MFCD19689724
:184.067 g/mol
Storage Temperature:Ambient
Frequently Bought Together