2-Methyl-3,5,6,7-tetrahydro-cyclopentapyrimidin-4-one 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:65818-01-1
:MFCD16872922
Formula:C₈H₁₀N₂O
MDL Number:MFCD08143584
:150.178 g/mol
Storage Temperature:Ambient
Frequently Bought Together