Order Entry
Netherlands
ContactUsLinkComponent
N-Cyclopropyl-1H-imidazole-1-carboxamide 95%
N-Cyclopropyl-1H-imidazole-1-carboxamide 95%
Catalog # APOSOR917028-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Cyclopropyl-1H-imidazole-1-carboxamide 95%
Catalog # APOSOR917028-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :151.166 g/mol
Storage Temperature:Ambient
CAS Number:1033090-34-4
Formula:C₇H₉N₃O
MDL Number:MFCD11099699