
3-(Fur-2-yl)-1-methyl-1H-pyrazole-5-carboxylic acid 97%
Catalog # APOSOR9148-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:192.171 g/mol
Melting Pt:130 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:859851-00-6
Formula:C₉H₈N₂O₃
MDL Number:MFCD08271950