N-Methyl-N-[3-(piperidin-1-ylmethyl)benzyl]amine 97%
Catalog # APOSOR9135-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:218.338 g/mol
Storage Temperature:Ambient
CAS Number:868755-58-2
Formula:C₁₄H₂₂N₂
MDL Number:MFCD08060527