Order Entry
Netherlands
Orders LinkContactUsLinkComponent
2-Methoxy-3-nitrobenzonitrile 98%

2-Methoxy-3-nitrobenzonitrile 98%

Supplier:   Apollo Scientific
2-Methoxy-3-nitrobenzonitrile 98%
Supplier:  Apollo Scientific
Certificates
2-Methoxy-3-nitrobenzonitrile 98%

2 Options Available Below

Product Details & Documents
  • CAS Number:121001-06-7
  • Formula:C₈H₆N₂O₃
  • MDL Number:MFCD18398370
  • Molecular Weight:178.145 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 220,00 €
    220,00 €
    1 * 1 g
       220,00 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 652,00 €
    652,00 €
    1 * 5 g
       652,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.