Order Entry
Netherlands
ContactUsLinkComponent
(3R)-1-Methylpiperidin-3-amine dihydrochloride 98%
(3R)-1-Methylpiperidin-3-amine dihydrochloride 98%
Catalog # APOSOR908084-1G
Supplier:  Apollo Scientific
CAS Number:  
(3R)-1-Methylpiperidin-3-amine dihydrochloride 98%
Catalog # APOSOR908084-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :187.111 g/mol
Storage Temperature:Ambient
CAS Number:1157849-50-7
Formula:C₆H₁₄N₂·2ClH
MDL Number:MFCD20926141