Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1-Benzothiophen-4-yl)boronic acid
(1-Benzothiophen-4-yl)boronic acid 98%
(1-Benzothiophen-4-yl)boronic acid 98%
APOSOR907505-1G
:
Apollo Scientific
:
(1-Benzothiophen-4-yl)boronic acid 98%
APOSOR907505-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
178.016 g/mol
:
Ambient
:
177735-30-7
:
C₈H₇BO₂S
:
MFCD22191951