
2-Bromo-1-(toluene-4-sulphonyl)-1H-pyrrole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:290306-56-8
:
Formula:C₁₁H₁₀BrNO₂S
MDL Number:MFCD07364061
:300.172 g/mol
Melting Pt:102…105 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together