Order Entry
Netherlands
ContactUsLinkComponent
2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine 95+%
Irritant
2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine 95+%
Catalog # APOSOR7807-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine 95+%
Catalog # APOSOR7807-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Melting Pt:87…89 °C
 :231.254 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:849925-00-4
Formula:C₁₁H₁₃N₅O
MDL Number:MFCD03426077