Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-(3-hydroxyprop-1-yn-1-yl)benzoate
Methyl 3-(3-hydroxyprop-1-yn-1-yl)benzoate
Methyl 3-(3-hydroxyprop-1-yn-1-yl)benzoate
Methyl 3-(3-hydroxyprop-1-yn-1-yl)benzoate
APOSOR7568-250MG
:
Apollo Scientific
:
Methyl 3-(3-hydroxyprop-1-yn-1-yl)benzoate
APOSOR7568-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
190.195 g/mol
:
GHS07
:
Ambient
:
121114-45-2
:
C₁₁H₁₀O₃
:
MFCD04039139