
5-Chloronaphthalene-1-sulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6291-07-2
:
Formula:C₁₀H₆Cl₂O₂S
MDL Number:MFCD00269949
:261.124 g/mol
Melting Pt:94…95 °C
Storage Temperature:Freezer
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together