
Methyl 4-(chlorosulphonyl)-3-methoxybenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:162141-56-2
:
Formula:C₉H₉ClO₅S
MDL Number:MFCD27959393
:264.683 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together