Order Entry
Netherlands
ContactUsLinkComponent
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydropyridine-1(2h)-carboxylate
Irritant
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydropyridine-1(2h)-carboxylate
Catalog # APOSOR55284-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydropyridine-1(2h)-carboxylate
Catalog # APOSOR55284-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :309.209 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1121057-77-9
Formula:C₁₆H₂₈BNO₄
MDL Number:MFCD18383341