1H-Imidazol-1-amine hydrochloride 97%
Catalog # APOSOR52986-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:119.553 g/mol
Storage Temperature:Ambient
CAS Number:83279-44-1
Formula:C₃H₅N₃·ClH
MDL Number:MFCD26390998