
Methyl 4-bromo-3-iodobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1257231-51-8
:
Formula:C₈H₆BrIO₂
MDL Number:MFCD18398802
:340.941 g/mol
Melting Pt:70…71 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together