Order Entry
Netherlands
ContactUsLinkComponent
3-(1-Methyl-1H-pyrazol-3-yl)benzoic acid 97%
Irritant
3-(1-Methyl-1H-pyrazol-3-yl)benzoic acid 97%
Catalog # APOSOR5217-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(1-Methyl-1H-pyrazol-3-yl)benzoic acid 97%
Catalog # APOSOR5217-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :202.209 g/mol
Melting Pt:167…169 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:906352-85-0
Formula:C₁₁H₁₀N₂O₂
MDL Number:MFCD09065006