
1-(2-Hydroxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1040377-08-9
:
Formula:C₁₁H₁₉BN₂O₃
MDL Number:MFCD12033564
:238.091 g/mol
:GHS07
Storage Temperature:Freezer
Frequently Bought Together