Order Entry
Netherlands
ContactUsLinkComponent
5-Mercapto-2-methoxy-4-methylbenzoic acid ≥98%
5-Mercapto-2-methoxy-4-methylbenzoic acid ≥98%
Catalog # APOSOR510234-250MG
Supplier:  Apollo Scientific
CAS Number:  
5-Mercapto-2-methoxy-4-methylbenzoic acid ≥98%
Catalog # APOSOR510234-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :198.239 g/mol
Storage Temperature:Ambient
CAS Number:439579-12-1
Formula:C₉H₁₀O₃S
MDL Number:MFCD09831918