Order Entry
Netherlands
ContactUsLinkComponent
Methyl 3-bromo-4-methoxy-2-methylbenzoate ≥98%
Methyl 3-bromo-4-methoxy-2-methylbenzoate ≥98%
Catalog # APOSOR510232-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-bromo-4-methoxy-2-methylbenzoate ≥98%
Catalog # APOSOR510232-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :259.097 g/mol
Storage Temperature:Ambient
CAS Number:1427501-73-2
Formula:C₁₀H₁₁BrO₃
MDL Number:MFCD23703479