Order Entry
Netherlands
ContactUsLinkComponent
2-Chloro-7-cyclopentyl-7H-pyrrolo[2,3-d]pyrimidine-6-carboxaldehyde ≥98%
2-Chloro-7-cyclopentyl-7H-pyrrolo[2,3-d]pyrimidine-6-carboxaldehyde ≥98%
Catalog # APOSOR510188-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-7-cyclopentyl-7H-pyrrolo[2,3-d]pyrimidine-6-carboxaldehyde ≥98%
Catalog # APOSOR510188-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :249.696 g/mol
Storage Temperature:Ambient
CAS Number:1211443-55-8
Formula:C₁₂H₁₂ClN₃O