Order Entry
Netherlands
ContactUsLinkComponent
Methyl 2-cyano-5-nitrobenzoate ≥97%
Methyl 2-cyano-5-nitrobenzoate ≥97%
Catalog # APOSOR510096-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-cyano-5-nitrobenzoate ≥97%
Catalog # APOSOR510096-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :206.155 g/mol
Storage Temperature:Refrigerator
CAS Number:1260777-93-2
Formula:C₉H₆N₂O₄
MDL Number:MFCD11975694