Order Entry
Netherlands
ContactUsLinkComponent
2-Chloro-6-methyl-4-nitropyridine > 98%
2-Chloro-6-methyl-4-nitropyridine > 98%
Catalog # APOSOR510059-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-6-methyl-4-nitropyridine > 98%
Catalog # APOSOR510059-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :172.569 g/mol
Storage Temperature:Freezer
CAS Number:79055-51-9
Formula:C₆H₅ClN₂O₂
MDL Number:MFCD14582081