Order Entry
Netherlands
ContactUsLinkComponent
3-Methyl-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride ≥98%
3-Methyl-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride ≥98%
Catalog # APOSOR510015-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-Methyl-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride ≥98%
Catalog # APOSOR510015-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :173.643 g/mol
Storage Temperature:Ambient
CAS Number:601515-50-8
Formula:C₇H₁₁N₃·ClH
MDL Number:MFCD15072031