Order Entry
Netherlands
ContactUsLinkComponent
 
(2R)-1,4-Dioxane-2-methanol 97%
Catalog # APOSOR49072-1G
Supplier:  Apollo Scientific
CAS Number:  
(2R)-1,4-Dioxane-2-methanol 97%
Catalog # APOSOR49072-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :118.131 g/mol
Storage Temperature:Refrigerator
CAS Number:406913-88-0
Formula:C₅H₁₀O₃
MDL Number:MFCD16620507