Order Entry
Netherlands
ContactUsLinkComponent
Methyl 2-bromo-4-(bromomethyl)benzoate 97%
Methyl 2-bromo-4-(bromomethyl)benzoate 97%
Catalog # APOSOR480825-5G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-bromo-4-(bromomethyl)benzoate 97%
Catalog # APOSOR480825-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :307.967 g/mol
Storage Temperature:Ambient
CAS Number:128577-48-0
Formula:C₉H₈Br₂O₂
MDL Number:MFCD03840738