Order Entry
Netherlands
ContactUsLinkComponent
(2S)-1-(1H-1,2,4-Triazol-1-yl)propan-2-ol 95%
(2S)-1-(1H-1,2,4-Triazol-1-yl)propan-2-ol 95%
Catalog # APOSOR480291-1G
Supplier:  Apollo Scientific
CAS Number:  
(2S)-1-(1H-1,2,4-Triazol-1-yl)propan-2-ol 95%
Catalog # APOSOR480291-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :127.144 g/mol
Storage Temperature:Ambient
Formula:C₅H₉N₃O
MDL Number:MFCD26042662