Order Entry
Netherlands
ContactUsLinkComponent
N-Methyl-3-phenyl-indan-1-amine hydrochloride
N-Methyl-3-phenyl-indan-1-amine hydrochloride
Catalog # APOSOR475153-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Methyl-3-phenyl-indan-1-amine hydrochloride
Catalog # APOSOR475153-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.774 g/mol
Storage Temperature:Ambient
Formula:C₁₆H₁₇N·ClH
MDL Number:MFCD31695628