Order Entry
Netherlands
ContactUsLinkComponent
4-Bromo-N-(1-phenylethyl)aniline hydrochloride
4-Bromo-N-(1-phenylethyl)aniline hydrochloride
Catalog # APOSOR475149-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-N-(1-phenylethyl)aniline hydrochloride
Catalog # APOSOR475149-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :312.633 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₄BrN·ClH
MDL Number:MFCD31695759