Order Entry
Netherlands
ContactUsLinkComponent
Ethyl 3-[(1-phenylethylamino)methyl]benzoate hydrochloride
Ethyl 3-[(1-phenylethylamino)methyl]benzoate hydrochloride
Catalog # APOSOR475112-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 3-[(1-phenylethylamino)methyl]benzoate hydrochloride
Catalog # APOSOR475112-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :319.826 g/mol
Storage Temperature:Ambient
Formula:C₁₈H₂₁NO₂·ClH
MDL Number:MFCD31695655