Ethyl 2-[[[(1S)-1-(4-bromophenyl)ethyl]amino]methyl]benzoate hydrochloride
Catalog # APOSOR475057-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Verpakking:Bottle
- VPE:1 g
- Seq:3047
Specifications
About this item
:
:398.722 g/mol
Storage Temperature:Kamertemperatuur
Formula:C₁₈H₂₀BrNO₂·ClH
MDL Number:MFCD31695639