Order Entry
Netherlands
ContactUsLinkComponent
 
Ethyl 4-[[[(1S)-1-(4-bromophenyl)ethyl]amino]methyl]benzoate hydrochloride
Catalog # APOSOR475055-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-[[[(1S)-1-(4-bromophenyl)ethyl]amino]methyl]benzoate hydrochloride
Catalog # APOSOR475055-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :398.722 g/mol
Storage Temperature:Ambient
Formula:C₁₈H₂₀BrNO₂·ClH
MDL Number:MFCD31695633