Order Entry
Netherlands
ContactUsLinkComponent
N-[(1S)-1-(4-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Irritant
N-[(1S)-1-(4-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Catalog # APOSOR475044-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-[(1S)-1-(4-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Catalog # APOSOR475044-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :274.585 g/mol
 :GHS07
Storage Temperature:Ambient
Formula:C₁₁H₁₂BrN·ClH
MDL Number:MFCD31695742