Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-3-(3-Bromophenyl)-2-hydroxypropionic acid
(R)-3-(3-Bromophenyl)-2-hydroxypropionic acid
(R)-3-(3-Bromophenyl)-2-hydroxypropionic acid
(R)-3-(3-Bromophenyl)-2-hydroxypropionic acid
APOSOR471448-5G
:
Apollo Scientific
:
(R)-3-(3-Bromophenyl)-2-hydroxypropionic acid
APOSOR471448-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
245.07 g/mol
:
Ambient
:
1932233-01-6
:
C₉H₉BrO₃
:
MFCD28977373