(R)-2-(2-Chlorophenyl)-2-hydroxypropionic acid
Catalog # APOSOR471443-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:200.619 g/mol
Storage Temperature:Ambient
CAS Number:1215178-13-4
Formula:C₉H₉ClO₃
MDL Number:MFCD05663617