Order Entry
Netherlands
ContactUsLinkComponent
(S)-2-(2-Chlorophenyl)-2-hydroxypropionic acid
(S)-2-(2-Chlorophenyl)-2-hydroxypropionic acid
Catalog # APOSOR471442-1G
Supplier:  Apollo Scientific
CAS Number:  
(S)-2-(2-Chlorophenyl)-2-hydroxypropionic acid
Catalog # APOSOR471442-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.619 g/mol
Storage Temperature:Ambient
CAS Number:1215178-12-3
Formula:C₉H₉ClO₃
MDL Number:MFCD05663616