Order Entry
Netherlands
ContactUsLinkComponent
3-[(4-Nitrophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
3-[(4-Nitrophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Catalog # APOSOR471400-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[(4-Nitrophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Catalog # APOSOR471400-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :348.307 g/mol
Storage Temperature:Ambient
CAS Number:2006277-51-4
Formula:C₁₆H₁₆N₂O₇
MDL Number:MFCD28977394