Methyl 3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylate
Catalog # APOSOR471399-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:278.218 g/mol
Storage Temperature:Ambient
CAS Number:2006277-45-6
Formula:C₁₂H₁₀N₂O₆
MDL Number:MFCD28977395