2-(Chloromethyl)-6-methoxyquinazoline
Catalog # APOSOR471368-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Verpakking:Bottle
- VPE:1 g
- Seq:3047
Specifications
About this item
:
:208.644 g/mol
Storage Temperature:Kamertemperatuur
CAS Number:1936095-76-9
Formula:C₁₀H₉ClN₂O
MDL Number:MFCD28977359