5-Methoxy-1-methyl-1,2,3,6-tetrahydropyridine
Catalog # APOSOR471143-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:127.184 g/mol
Storage Temperature:Ambient
CAS Number:98435-42-8
Formula:C₇H₁₃NO
MDL Number:MFCD13705327