Order Entry
Netherlands
ContactUsLinkComponent
N-Boc-3-(4-Pyridyl)-2-propyn-1-amine
N-Boc-3-(4-Pyridyl)-2-propyn-1-amine
Catalog # APOSOR471132-5G
Supplier:  Apollo Scientific
CAS Number:  
N-Boc-3-(4-Pyridyl)-2-propyn-1-amine
Catalog # APOSOR471132-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :232.278 g/mol
Storage Temperature:Ambient
CAS Number:777856-67-4
Formula:C₁₃H₁₆N₂O₂
MDL Number:MFCD24447935