

1-Bromo-3-hydroxynaphthalene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5498-31-7
:
Formula:C₁₀H₇BrO
MDL Number:MFCD01712260
:223.066 g/mol
Boiling Pt:130…132 °C (1 mm Hg)
Melting Pt:117…120 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together