1-(4-Iodophenyl)-3-morpholino-5,6-dihydropyridin-2(1H)-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:473927-69-4
:
Formula:C₁₅H₁₇IN₂O₂
MDL Number:MFCD18072445
:384.212 g/mol
Storage Temperature:Ambient
Frequently Bought Together