Order Entry
Netherlands
ContactUsLinkComponent
1-(3-Chloropropyl)imidazole
1-(3-Chloropropyl)imidazole
Catalog # APOSOR471035-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3-Chloropropyl)imidazole
Catalog # APOSOR471035-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :144.602 g/mol
Storage Temperature:Ambient
CAS Number:53710-78-4
Formula:C₆H₉ClN₂
MDL Number:MFCD09728547