4-(Chloromethyl)piperidine hydrochloride
Catalog # APOSOR471034-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:170.08 g/mol
Storage Temperature:Ambient
CAS Number:1822-61-3
Formula:C₆H₁₂ClN·ClH
MDL Number:MFCD11110948