Methyl 2-[Imino(methoxy)methyl]benzoate hydrochloride
Catalog # APOSOR470982-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:229.66 g/mol
Storage Temperature:Ambient
CAS Number:1823954-95-5
Formula:C₁₀H₁₁NO₃·ClH
MDL Number:MFCD28014553