Order Entry
Netherlands
ContactUsLinkComponent
Methyl 3-[Imino(methoxy)methyl]benzoate hydrochloride
Methyl 3-[Imino(methoxy)methyl]benzoate hydrochloride
Catalog # APOSOR470981-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-[Imino(methoxy)methyl]benzoate hydrochloride
Catalog # APOSOR470981-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.66 g/mol
Storage Temperature:Ambient
CAS Number:1154063-16-7
Formula:C₁₀H₁₁NO₃·ClH
MDL Number:MFCD10697976